This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02659859_m1
See other NXPE4 GE Assays ›
Gene Symbol:
NXPE4
Gene Name:
neurexophilin and PC-esterase domain family member 4
Gene Aliases:
FAM55D
Chromosome Location:
Chr.5: 18675387 - 18724566 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02659859_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NXPE4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005619744.2 XP_005619801.1 1367 128
XM_022418283.1 XP_022273991.1 1118 128
XM_022418284.1 XP_022273992.1 1237 128
XM_022418285.1 XP_022273993.1 1099 128
XM_022418286.1 XP_022273994.1 1074 128
XM_022418287.1 XP_022273995.1 1192 128
XM_022418288.1 XP_022273996.1 1055 128
XM_022418289.1 XP_022273997.1 1486 128
XM_022418290.1 XP_022273998.1 1309 128
XM_022418291.1 XP_022273999.1 864 128
XM_022418293.1 XP_022274001.1 1412 128
XM_022418294.1 XP_022274002.1 1325 128
XM_022418295.1 XP_022274003.1 1264 128
XM_022418296.1 XP_022274004.1 1206 128
XM_022418297.1 XP_022274005.1 1428 128

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Canis familiaris

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0