This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02659870_m1
See other DRAM1 GE Assays ›
Gene Symbol:
DRAM1
Gene Name:
DNA damage regulated autophagy modulator 1
Gene Aliases:
-
Chromosome Location:
Chr.15: 40759739 - 40794666 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02659870_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DRAM1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_022403421.1 XP_022259129.1 4 - 5 520 69
XM_022403422.1 XP_022259130.1 4 - 5 520 69
XM_022403423.1 XP_022259131.1 4 - 5 520 69
XM_022403424.1 XP_022259132.1 4 - 5 520 69
XM_022403425.1 XP_022259133.1 3 - 4 421 69
XM_022403426.1 XP_022259134.1 4 - 5 520 69
XM_022403427.1 XP_022259135.1 3 - 4 421 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Canis familiaris Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0