This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02660056_m1
See other MOB2 GE Assays ›
Gene Symbol:
MOB2
Gene Name:
MOB kinase activator 2
Gene Aliases:
-
Chromosome Location:
Chr.18: 45698096 - 45776402 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02660056_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MOB2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005631331.3 XP_005631388.1 3 - 4 365 82
XM_022405516.1 XP_022261224.1 3 - 4 275 82
XM_022405517.1 XP_022261225.1 4 - 5 447 82
XM_849165.5 XP_854258.2 3 - 4 403 82

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

kinase activator

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-cj8gb:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline