This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02660559_m1
See other ZBP1 GE Assays ›
Gene Symbol:
ZBP1
Gene Name:
Z-DNA binding protein 1
Gene Aliases:
-
Chromosome Location:
Chr.24: 42583674 - 42589857 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02660559_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ZBP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_014107314.2 XP_013962789.1 2 - 3 2900 56
XM_014107315.2 XP_013962790.1 2 - 3 2897 56
XM_014107316.2 XP_013962791.1 3 - 4 569 56

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

DNA metabolism protein nucleic acid metabolism protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0