This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02664045_m1
See other NUDT5 GE Assays ›
Gene Symbol:
NUDT5
Gene Name:
nudix hydrolase 5
Gene Aliases:
-
Chromosome Location:
Chr.2: 24239632 - 24266763 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02664045_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NUDT5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_014122641.2 XP_013978116.1 492 62
XM_022407815.1 XP_022263523.1 355 62
XM_022407824.1 XP_022263532.1 473 62
XM_022407830.1 XP_022263538.1 473 62
XM_022407835.1 XP_022263543.1 572 62
XM_535190.6 XP_535190.2 515 62

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Canis familiaris

Panther Classification:

Molecular Function -

pyrophosphatase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0