This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02664661_g1
See other ATP5PO GE Assays ›
Gene Symbol:
ATP5PO ATP5O
Gene Name:
ATP synthase peripheral stalk subunit OSCP
ATP synthase, H+ transporting, mitochondrial F1 complex, O subunit
Gene Aliases:
ATP5O
ATP50
Chromosome Location:
Chr.31: 29370861 - 29380512 on Build CanFam3.1
Chromosome Location:
Chr.31: 28472106 - 28481759 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
-
Assay ID Cf02664661_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ATP5PO
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005638881.3 XP_005638938.1 6 - 7 623 129
Gene Symbol: ATP5O
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005638862.3 XP_005638919.1 6 - 7 623 129
GenBank mRNA CV999268.1 - 6 - 7 545 129

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

ATP synthase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-pc55z:80/100.66.76.57:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0