This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02668256_gH
See other DBI GE Assays ›
Gene Symbol:
DBI
Gene Name:
diazepam binding inhibitor, acyl-CoA binding protein
Gene Aliases:
ACBP, EP
Chromosome Location:
Chr.19: 30640429 - 30645704 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02668256_gH
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DBI
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001197047.1 NP_001183976.1 2 - 3 205 134
XM_014121370.2 XP_013976845.1 2 - 3 258 134

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Canis familiaris Inventoried Probe spans exons

Panther Classification:

Molecular Function -

transfer/carrier protein

Gene Ontology Categories:

Process(es)

fatty-acyl-CoA binding
lipid binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-gjvvj:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0