This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02672730_u1
See other TMEM25 GE Assays ›
Gene Symbol:
TMEM25
Gene Name:
transmembrane protein 25
Gene Aliases:
-
Chromosome Location:
Chr.5: 15202726 - 15207252 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02672730_u1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TMEM25
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005619710.3 XP_005619767.1 948 127
XM_022418242.1 XP_022273950.1 661 127
XM_848285.4 XP_853378.1 949 127

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline