Assay Details
| Gene Symbol: ATF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014115083.2 | XP_013970558.1 | 1088 | 183 | ||
| XM_022420880.1 | XP_022276588.1 | 1142 | 183 | |||
| XM_847382.5 | XP_852475.1 | 420 | 183 | |||
More Information
Set Membership: |
|

ATF3
Genomic Map