This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02676513_s1
See other SIAH1 GE Assays ›
Gene Symbol:
SIAH1
Gene Name:
siah E3 ubiquitin protein ligase 1
Gene Aliases:
-
Chromosome Location:
Chr.2: 66630795 - 66709497 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02676513_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SIAH1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003638931.4 XP_003638979.1 2 1269 151
XM_005617594.3 XP_005617651.1 2 848 151
XM_014109624.2 XP_013965099.1 6 1462 151
XM_014109629.2 XP_013965104.1 7 1613 151
XM_022411602.1 XP_022267310.1 7 1583 151
XM_022411606.1 XP_022267314.1 7 1580 151
XM_022411610.1 XP_022267318.1 6 1346 151
XM_022411612.1 XP_022267320.1 5 1221 151
XM_022411622.1 XP_022267330.1 2 738 151

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

ubiquitin-protein ligase protein modifying enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-f5rhb:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0