This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02678953_m1
See other HMGCLL1 GE Assays ›
Gene Symbol:
HMGCLL1
Gene Name:
3-hydroxymethyl-3-methylglutaryl-CoA lyase like 1
Gene Aliases:
-
Chromosome Location:
Chr.12: 22748798 - 22953849 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02678953_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HMGCLL1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005627493.3 XP_005627550.2 3 - 4 397 153
XM_014118065.2 XP_013973540.1 3 - 4 288 153
XM_014118066.2 XP_013973541.1 3 - 4 213 153
XM_022426167.1 XP_022281875.1 3 - 4 398 153
XM_538973.6 XP_538973.4 3 - 4 394 153

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
This assay is in the 5'UTR region of the XM_014118065.2, XM_014118066.2 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

lyase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hldrw:80/100.66.76.65:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0