This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02679462_g1
See other MAMSTR GE Assays ›
Gene Symbol:
MAMSTR
Gene Name:
MEF2 activating motif and SAP domain containing transcriptional regulator
Gene Aliases:
-
Chromosome Location:
Chr.1: 107598273 - 107605481 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02679462_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MAMSTR
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005616338.3 XP_005616395.1 6 - 7 850 112
XM_005616340.3 XP_005616397.1 7 - 8 947 112
XM_005616341.3 XP_005616398.1 7 - 8 889 112
XM_014119090.2 XP_013974565.1 6 - 7 2399 112
XM_014119092.2 XP_013974567.1 4 - 5 2094 112
XM_014119095.2 XP_013974570.1 5 - 6 2182 112
XM_014119100.2 XP_013974575.1 6 - 7 2219 112
XM_014119101.2 XP_013974576.1 7 - 8 1126 112
XM_022424159.1 XP_022279867.1 5 - 6 2121 112

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hvxbk:80/100.66.77.65:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0