This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02680225_m1
See other HDAC11 GE Assays ›
Gene Symbol:
HDAC11
Gene Name:
histone deacetylase 11
Gene Aliases:
-
Chromosome Location:
Chr.20: 3792832 - 3815922 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02680225_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HDAC11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005632106.2 XP_005632163.1 699 90
XM_005632107.3 XP_005632164.1 721 90
XM_022406335.1 XP_022262043.1 785 90
XM_022406336.1 XP_022262044.1 1051 90
XM_022406337.1 XP_022262045.1 1246 90
XM_022406338.1 XP_022262046.1 771 90

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Canis familiaris

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline