This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02680880_u1
See other HSD11B1L GE Assays ›
Gene Symbol:
HSD11B1L
Gene Name:
hydroxysteroid 11-beta dehydrogenase 1 like
Gene Aliases:
-
Chromosome Location:
Chr.20: 54336451 - 54341738 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02680880_u1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HSD11B1L
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005633011.3 XP_005633068.1 7 810 133
XM_005633012.2 XP_005633069.1 7 701 133
XM_014122336.2 XP_013977811.1 7 1671 133
XM_014122337.2 XP_013977812.1 7 1002 133
XM_014122338.2 XP_013977813.1 7 1669 133
XM_014122339.2 XP_013977814.1 6 2245 133
XM_014122340.2 XP_013977815.1 7 806 133
XM_014122341.2 XP_013977816.1 5 2130 133
XM_022407178.1 XP_022262886.1 7 1833 133
XM_022407179.1 XP_022262887.1 7 1861 133
XM_022407180.1 XP_022262888.1 7 1563 133
XM_022407181.1 XP_022262889.1 7 1673 133
XM_022407182.1 XP_022262890.1 7 697 133

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris Amplicon spans exons and probe does not span exons

Panther Classification:

Molecular Function -

dehydrogenase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0