This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02681531_s1
See other NXPE4 GE Assays ›
Gene Symbol:
NXPE4
Gene Name:
neurexophilin and PC-esterase domain family member 4
Gene Aliases:
FAM55D
Chromosome Location:
Chr.5: 18675387 - 18724566 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02681531_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NXPE4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005619744.2 XP_005619801.1 1891 83
XM_022418283.1 XP_022273991.1 1642 83
XM_022418284.1 XP_022273992.1 1761 83
XM_022418285.1 XP_022273993.1 1623 83
XM_022418286.1 XP_022273994.1 1598 83
XM_022418287.1 XP_022273995.1 1716 83
XM_022418288.1 XP_022273996.1 1579 83
XM_022418289.1 XP_022273997.1 2010 83
XM_022418290.1 XP_022273998.1 1833 83
XM_022418291.1 XP_022273999.1 1388 83
XM_022418293.1 XP_022274001.1 1936 83
XM_022418294.1 XP_022274002.1 1849 83
XM_022418295.1 XP_022274003.1 1788 83
XM_022418296.1 XP_022274004.1 1730 83
XM_022418297.1 XP_022274005.1 1745 83

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Canis familiaris

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0