This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02683425_u1
See other MTHFD2 GE Assays ›
Gene Symbol:
MTHFD2
Gene Name:
methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2, methenyltetrahydrofolate cyclohydrolase
Gene Aliases:
-
Chromosome Location:
Chr.17: 48882838 - 48896816 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02683425_u1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MTHFD2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005630551.3 XP_005630608.1 2 318 154

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Canis familiaris Amplicon spans exons and probe does not span exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-794c4b9cd6-65nqn:80/100.66.78.26:80.
git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.52.0-Offline