This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02684815_m1
See other ELAVL2 GE Assays ›
Gene Symbol:
ELAVL2
Gene Name:
ELAV like RNA binding protein 2
Gene Aliases:
-
Chromosome Location:
Chr.11: 42504205 - 42647093 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02684815_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ELAVL2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005626684.3 XP_005626741.1 4 - 5 989 133
XM_014117792.2 XP_013973267.1 4 - 5 1014 133
XM_014117793.2 XP_013973268.1 4 - 5 538 133
XM_014117794.2 XP_013973269.1 4 - 5 514 133
XM_014117795.2 XP_013973270.1 3 - 4 898 133
XM_014117796.2 XP_013973271.1 3 - 4 443 133
XM_022425480.1 XP_022281188.1 4 - 5 1014 133
XM_022425481.1 XP_022281189.1 4 - 5 989 133
XM_022425482.1 XP_022281190.1 4 - 5 987 133
XM_022425483.1 XP_022281191.1 4 - 5 986 133
XM_022425484.1 XP_022281192.1 4 - 5 1014 133
XM_022425485.1 XP_022281193.1 4 - 5 1014 133
XM_022425486.1 XP_022281194.1 4 - 5 989 133
XM_022425487.1 XP_022281195.1 4 - 5 989 133
XM_022425488.1 XP_022281196.1 3 - 4 895 133

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

translation initiation factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0