This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02692499_m1
See other RBPMS GE Assays ›
Gene Symbol:
RBPMS
Gene Name:
RNA binding protein, mRNA processing factor
Gene Aliases:
-
Chromosome Location:
Chr.16: 33788675 - 33961595 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02692499_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RBPMS
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005629903.3 XP_005629960.1 566 97
XM_014120008.2 XP_013975483.1 879 97
XM_014120014.2 XP_013975489.1 491 97
XM_014120016.2 XP_013975491.1 879 97
XM_022404088.1 XP_022259796.1 879 97
XM_022404089.1 XP_022259797.1 609 97
XM_022404090.1 XP_022259798.1 879 97
XM_022404091.1 XP_022259799.1 879 97
XM_022404092.1 XP_022259800.1 879 97
XM_022404093.1 XP_022259801.1 879 97
XM_022404094.1 XP_022259802.1 265 97
XM_022404095.1 XP_022259803.1 816 97
XM_022404097.1 XP_022259805.1 879 97
XM_022404098.1 XP_022259806.1 879 97
XM_022404099.1 XP_022259807.1 220 97
XM_022404100.1 XP_022259808.1 277 97
XM_022404101.1 XP_022259809.1 879 97
XM_022404102.1 XP_022259810.1 503 97
XM_022404104.1 XP_022259812.1 879 97
XM_022404105.1 XP_022259813.1 150 97

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_022404105.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Canis familiaris

Panther Classification:

Molecular Function -

RNA splicing factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline