This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02694221_m1
See other DUSP15 GE Assays ›
Gene Symbol:
DUSP15
Gene Name:
dual specificity phosphatase 15
Gene Aliases:
-
Chromosome Location:
Chr.24: 21323751 - 21332584 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02694221_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DUSP15
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005634917.3 XP_005634974.1 4 - 5 188 147
XM_005634918.3 XP_005634975.1 4 - 5 260 147
XM_847171.5 XP_852264.2 4 - 5 306 147

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_005634918.3 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

protein phosphatase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline