This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02698334_g1
See other LIPE GE Assays ›
Gene Symbol:
LIPE
Gene Name:
lipase E, hormone sensitive type
Gene Aliases:
HSL
Chromosome Location:
Chr.1: 111987862 - 112006492 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02698334_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LIPE
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005616535.3 XP_005616592.1 5 - 6 2116 166
XM_005616536.3 XP_005616593.1 5 - 6 1136 166
XM_005616537.3 XP_005616594.1 5 - 6 1168 166
XM_005616538.3 XP_005616595.1 5 - 6 1102 166
XM_005616539.3 XP_005616596.1 5 - 6 1273 166
XM_533655.7 XP_533655.4 5 - 6 2051 166
GenBank mRNA AY063230.1 - 4 - 5 440 166

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

lipase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0