This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02698990_m1
See other TFDP2 GE Assays ›
Gene Symbol:
TFDP2
Gene Name:
transcription factor Dp-2
Gene Aliases:
-
Chromosome Location:
Chr.23: 37757858 - 37871632 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02698990_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TFDP2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005634531.3 XP_005634588.1 3 - 4 305 109
XM_005634532.3 XP_005634589.1 3 - 4 305 109
XM_022408708.1 XP_022264416.1 2 - 3 303 109
XM_534293.6 XP_534293.2 2 - 3 152 109
XM_847279.5 XP_852372.3 3 - 4 305 109

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_534293.6 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

DNA-binding transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4g9kn:80/100.66.78.166:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0