This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02699846_m1
See other BRCA1 GE Assays ›
Gene Symbol:
BRCA1
Gene Name:
BRCA1 DNA repair associated
Gene Aliases:
-
Chromosome Location:
Chr.9: 19958941 - 20025494 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02699846_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BRCA1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001013416.1 NP_001013434.1 5204 150
XM_005624312.2 XP_005624369.1 5367 150
XM_005624313.3 XP_005624370.1 5486 150
XM_005624314.3 XP_005624371.1 5544 150
XM_014116455.1 XP_013971930.1 5395 150
XM_014116457.1 XP_013971932.1 5356 150
XM_014116458.1 XP_013971933.1 2065 150
XM_014116459.1 XP_013971934.1 2026 150
XM_022422510.1 XP_022278218.1 5446 150
XM_022422511.1 XP_022278219.1 5504 150
XM_022422512.1 XP_022278220.1 5328 150
XM_022422513.1 XP_022278221.1 5395 150
XM_022422514.1 XP_022278222.1 5154 150
GenBank mRNA AY136626.1 - 1214 150

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Canis familiaris

Panther Classification:

Molecular Function -

ubiquitin-protein ligase protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription from RNA polymerase II promoter
fatty acid biosynthetic process
cell cycle
negative regulation of fatty acid biosynthetic process
positive regulation of transcription, DNA-templated
protein autoubiquitination
protein K6-linked ubiquitination
DNA binding
transcription coactivator activity
ubiquitin-protein transferase activity
zinc ion binding
RNA polymerase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0