This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02703820_gH
See other IGFALS GE Assays ›
Gene Symbol:
IGFALS
Gene Name:
insulin like growth factor binding protein acid labile subunit
Gene Aliases:
-
Chromosome Location:
Chr.6: 39115744 - 39118831 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02703820_gH
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IGFALS
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_547189.6 XP_547189.5 2 1436 63

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jmd5k:80/100.66.78.41:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0