This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02703985_m1
See other SLCO1A2 GE Assays ›
Gene Symbol:
SLCO1A2
Gene Name:
solute carrier organic anion transporter family member 1A2
Gene Aliases:
OATPA
Chromosome Location:
Chr.27: 26279654 - 26398000 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02703985_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLCO1A2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005637072.3 XP_005637129.1 11 - 12 1647 105
XM_005637073.3 XP_005637130.1 14 - 15 2044 105
XM_005637075.3 XP_005637132.1 12 - 13 1895 105
XM_005637076.3 XP_005637133.1 12 - 13 1986 105
XM_014108664.2 XP_013964139.1 14 - 15 2135 105
XM_014108669.2 XP_013964144.1 14 - 15 2039 105
XM_022411207.1 XP_022266915.1 15 - 16 2084 105
XM_022411208.1 XP_022266916.1 13 - 14 1902 105
XM_022411209.1 XP_022266917.1 10 - 11 1505 105
XM_022411210.1 XP_022266918.1 13 - 14 1897 105
XM_022411211.1 XP_022266919.1 10 - 11 1486 105
XM_847746.5 XP_852839.2 11 - 12 1628 105

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris Inventoried Probe spans exons

Panther Classification:

Molecular Function -

transporter

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0