This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02704256_s1
See other GPR1 GE Assays ›
Gene Symbol:
GPR1
Gene Name:
G protein-coupled receptor 1
Gene Aliases:
-
Chromosome Location:
Chr.37: 14725573 - 14776009 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02704256_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPR1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005640575.3 XP_005640632.1 4 805 89
XM_005640576.3 XP_005640633.1 4 804 89
XM_005640578.3 XP_005640635.1 4 610 89
XM_005640581.3 XP_005640638.1 5 2041 89
XM_005640582.3 XP_005640639.1 3 727 89
XM_005640583.3 XP_005640640.1 3 729 89
XM_014111130.2 XP_013966605.1 5 938 89
XM_022415109.1 XP_022270817.1 5 685 89
XM_022415110.1 XP_022270818.1 5 930 89
XM_022415111.1 XP_022270819.1 4 623 89
XM_022415112.1 XP_022270820.1 4 673 89
XM_022415113.1 XP_022270821.1 4 609 89
XM_022415114.1 XP_022270822.1 4 970 89
XM_022415116.1 XP_022270824.1 5 2015 89
XM_022415117.1 XP_022270825.1 3 1839 89
XM_022415118.1 XP_022270826.1 3 597 89

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

G-protein coupled receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline