Assay Details
| Gene Symbol: B3GNT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005626144.2 | XP_005626201.1 | 2 | 1125 | 153 | |
| XM_531841.6 | XP_531841.1 | 2 | 1069 | 153 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

B3GNT2
Genomic Map