This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02708065_m1
See other HACD1 GE Assays ›
Gene Symbol:
HACD1
Gene Name:
3-hydroxyacyl-CoA dehydratase 1
Gene Aliases:
PTPLA
Chromosome Location:
Chr.2: 19362488 - 19383090 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02708065_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HACD1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001025269.1 NP_001020440.1 407 90
GenBank mRNA AJ876904.1 - 407 90

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length between 86 and 100 Recommended Assay Canis familiaris

Gene Ontology Categories:

Function(s) Process(es)

sphingolipid biosynthetic process
fatty acid elongation
negative regulation of phosphoprotein phosphatase activity
very long-chain fatty acid biosynthetic process
protein phosphatase inhibitor activity
3-hydroxyacyl-CoA dehydratase activity
enzyme binding
very-long-chain 3-hydroxyacyl-CoA dehydratase activity
3-hydroxy-arachidoyl-CoA dehydratase activity
3-hydroxy-behenoyl-CoA dehydratase activity
3-hydroxy-lignoceroyl-CoA dehydratase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0