This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02710657_u1
See other GDE1 GE Assays ›
Gene Symbol:
GDE1
Gene Name:
glycerophosphodiester phosphodiesterase 1
Gene Aliases:
-
Chromosome Location:
Chr.6: 25740741 - 25765404 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02710657_u1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GDE1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_022420035.1 XP_022275743.1 1 416 151
XM_022420036.1 XP_022275744.1 1 411 147
XM_536954.6 XP_536954.2 1 413 151

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
This assay is in the 5'UTR region of the XM_022420036.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Canis familiaris Amplicon spans exons and probe does not span exons

Panther Classification:

Molecular Function -

phosphodiesterase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline