This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02716346_g1
See other FGF1 GE Assays ›
Gene Symbol:
FGF1
Gene Name:
fibroblast growth factor 1
Gene Aliases:
ECGF, FGF-1, HBGF-1, aFGF
Chromosome Location:
Chr.2: 37467431 - 37554770 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02716346_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FGF1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_014108102.2 XP_013963577.1 287 77
XM_022408919.1 XP_022264627.1 170 77
XM_022408930.1 XP_022264638.1 562 77
XM_844181.5 XP_849274.1 416 77

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length between 71 and 85 Recommended Assay Canis familiaris

Panther Classification:

Molecular Function -

growth factor intercellular signal molecule

Gene Ontology Categories:

Function(s) Process(es)

activation of MAPK activity
angiogenesis
positive regulation of cell proliferation
fibroblast growth factor receptor signaling pathway
positive regulation of endothelial cell migration
cell differentiation
positive regulation of cell migration
activation of protein kinase B activity
cellular response to heat
positive regulation of cholesterol biosynthetic process
positive regulation of angiogenesis
positive regulation of transcription from RNA polymerase II promoter
positive regulation of cell division
branch elongation involved in ureteric bud branching
mesonephric epithelium development
regulation of endothelial tube morphogenesis
positive regulation of intracellular signal transduction
positive regulation of sprouting angiogenesis
fibroblast growth factor receptor binding
integrin binding
growth factor activity
heparin binding
S100 protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline