Assay Details
| Gene Symbol: SNX33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014109616.2 | XP_013965091.1 | 1 | 1735 | 148 | |
| XM_022412544.1 | XP_022268252.1 | 1 | 1735 | 148 | ||
| XM_022412545.1 | XP_022268253.1 | 1 | 1735 | 148 | ||
More Information
Set Membership: |
|

SNX33
Genomic Map