Assay Details
| Gene Symbol: ZNF454 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003431556.4 | XP_003431604.3 | 1007 | 196 | ||
| XM_014117614.2 | XP_013973089.1 | 982 | 196 | |||
More Information
Set Membership: |
|

ZNF454
Genomic Map