This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02723322_m1
See other NXPE4 GE Assays ›
Gene Symbol:
NXPE4
Gene Name:
neurexophilin and PC-esterase domain family member 4
Gene Aliases:
FAM55D
Chromosome Location:
Chr.5: 18675387 - 18724566 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02723322_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NXPE4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005619744.2 XP_005619801.1 1631 184
XM_022418283.1 XP_022273991.1 1382 184
XM_022418284.1 XP_022273992.1 1501 184
XM_022418285.1 XP_022273993.1 1363 184
XM_022418286.1 XP_022273994.1 1338 184
XM_022418287.1 XP_022273995.1 1456 184
XM_022418288.1 XP_022273996.1 1319 184
XM_022418289.1 XP_022273997.1 1750 184
XM_022418290.1 XP_022273998.1 1573 184
XM_022418291.1 XP_022273999.1 1128 184
XM_022418293.1 XP_022274001.1 1676 184
XM_022418294.1 XP_022274002.1 1589 184
XM_022418295.1 XP_022274003.1 1528 184
XM_022418296.1 XP_022274004.1 1470 184

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Canis familiaris

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0