This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02724698_m1
See other MIER1 GE Assays ›
Gene Symbol:
MIER1
Gene Name:
MIER1 transcriptional regulator
Gene Aliases:
-
Chromosome Location:
Chr.5: 43531847 - 43590102 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02724698_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIER1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005620186.3 XP_005620243.1 6 - 7 506 91
XM_005620192.3 XP_005620249.1 4 - 5 394 91
XM_014113678.2 XP_013969153.1 4 - 5 363 91
XM_014113680.2 XP_013969155.1 4 - 5 361 91
XM_022418680.1 XP_022274388.1 4 - 5 366 91
XM_022418681.1 XP_022274389.1 4 - 5 365 91
XM_022418682.1 XP_022274390.1 4 - 5 364 91
XM_022418683.1 XP_022274391.1 6 - 7 476 91
XM_022418684.1 XP_022274392.1 6 - 7 506 91
XM_022418685.1 XP_022274393.1 6 - 7 467 91
XM_022418686.1 XP_022274394.1 4 - 5 365 91
XM_022418687.1 XP_022274395.1 4 - 5 395 91
XM_022418688.1 XP_022274396.1 6 - 7 494 91
XM_022418689.1 XP_022274397.1 5 - 6 434 91
XM_022418690.1 XP_022274398.1 5 - 6 957 91
XM_022418691.1 XP_022274399.1 6 - 7 497 91
XM_022418692.1 XP_022274400.1 4 - 5 398 91
XM_022418693.1 XP_022274401.1 4 - 5 922 91
XM_022418694.1 XP_022274402.1 4 - 5 919 91
XM_022418695.1 XP_022274403.1 4 - 5 390 91
XM_022418696.1 XP_022274404.1 3 - 4 389 91
XM_022418697.1 XP_022274405.1 4 - 5 383 91
XM_022418698.1 XP_022274406.1 4 - 5 880 91
XM_022418700.1 XP_022274408.1 3 - 4 254 91
XM_022418701.1 XP_022274409.1 3 - 4 276 91
XM_846977.5 XP_852070.1 4 - 5 395 91

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length between 86 and 100 Recommended Assay Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

homeodomain transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0