This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02725480_m1
See other MSANTD2 GE Assays ›
Gene Symbol:
MSANTD2
Gene Name:
Myb/SANT DNA binding domain containing 2
Gene Aliases:
C5H11orf61
Chromosome Location:
Chr.5: 9623023 - 9654860 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02725480_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MSANTD2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_014113335.2 XP_013968810.1 194 122
XM_014113336.2 XP_013968811.1 734 122
XM_022418153.1 XP_022273861.1 1291 122
XM_844328.5 XP_849421.1 817 122
XM_853548.5 XP_858641.1 660 122

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Canis familiaris

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0