This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02730412_m1
See other UCP2 GE Assays ›
Gene Symbol:
UCP2
Gene Name:
uncoupling protein 2
Gene Aliases:
-
Chromosome Location:
Chr.21: 24304410 - 24311392 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02730412_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UCP2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001003048.1 NP_001003048.1 3 - 4 536 69
XM_005633268.2 XP_005633325.1 5 - 6 949 69
XM_022407413.1 XP_022263121.1 5 - 6 995 69
XM_022407414.1 XP_022263122.1 5 - 6 949 69
GenBank mRNA AB020887.1 - 3 - 4 536 69
AB611704.1 - 5 - 6 857 69
AF201377.1 - 3 - 4 528 69

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Canis familiaris Probe spans exons

Panther Classification:

Molecular Function -

transporter secondary carrier transporter

Gene Ontology Categories:

Function(s)

mitochondrial transport

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-9bd97d8-8bg8g:80/100.66.75.22:80.
git-commit: 653a2953ec6bb2e3dd1e37289b36fc4e0ab15bfe
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-Offline