This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02730440_m1
See other SEC22C GE Assays ›
Gene Symbol:
SEC22C
Gene Name:
SEC22 homolog C, vesicle trafficking protein
Gene Aliases:
-
Chromosome Location:
Chr.23: 11679576 - 11708047 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02730440_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SEC22C
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005634293.2 XP_005634350.1 4 - 5 691 132
XM_005634294.3 XP_005634351.1 4 - 5 834 132
XM_014106754.2 XP_013962229.1 4 - 5 847 132
XM_014106755.2 XP_013962230.1 5 - 6 670 132
XM_022408520.1 XP_022264228.1 4 - 5 729 132
XM_022408521.1 XP_022264229.1 4 - 5 608 132
XM_845320.5 XP_850413.1 4 - 5 687 132

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Canis familiaris Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0