Assay Details
| Gene Symbol: PCDH9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022408012.1 | XP_022263720.1 | 3 - 4 | 3817 | 101 | |
| XM_022408014.1 | XP_022263722.1 | 3 - 4 | 3862 | 101 | ||
| XM_843846.5 | XP_848939.1 | 3 - 4 | 3862 | 101 | ||
More Information
Set Membership: |
|

PCDH9
Genomic Map