This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf02739007_g1
See other GLUL GE Assays ›
Gene Symbol:
GLUL
Gene Name:
glutamate-ammonia ligase
Gene Aliases:
GS
Chromosome Location:
Chr.7: 15730898 - 15740241 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf02739007_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GLUL
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001002965.2 NP_001002965.2 1087 113
NM_001293224.1 NP_001280153.1 1 967 113
GenBank mRNA AF544242.1 - 1 1091 113
DN875727.1 - 1 329 113

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

ligase

Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
cell proliferation
cellular response to starvation
response to glucose
regulation of endothelial cell migration
protein palmitoylation
ribosome biogenesis
regulation of sprouting angiogenesis
regulation of protein localization to nucleolus
glutamate-ammonia ligase activity
ATP binding
protein-cysteine S-palmitoyltransferase activity
identical protein binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hwq2n:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0