Assay Details
| Gene Symbol: SLC25A41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014121928.2 | XP_013977403.1 | 3 | 1071 | 103 | |
| XM_014121929.2 | XP_013977404.1 | 3 | 1080 | 103 | ||
| XM_022407136.1 | XP_022262844.1 | 2 | 836 | 103 | ||
| XM_022407137.1 | XP_022262845.1 | 4 | 839 | 103 | ||
| XM_022407138.1 | XP_022262846.1 | 4 | 868 | 103 | ||
| XM_022407139.1 | XP_022262847.1 | 2 | 836 | 103 | ||
| XM_022407140.1 | XP_022262848.1 | 4 | 391 | 103 | ||
| XM_022407141.1 | XP_022262849.1 | 4 | 420 | 103 | ||
| XM_022407142.1 | XP_022262850.1 | 3 | 354 | 103 | ||
More Information
Set Membership: |
|

SLC25A41
Genomic Map