This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf03054097_m1
See other CEACAM1 GE Assays ›
Gene Symbol:
CEACAM1
Gene Name:
CEA cell adhesion molecule 1
Gene Aliases:
-
Chromosome Location:
Chr.1: 111902429 - 111919191 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf03054097_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CEACAM1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001097557.1 NP_001091026.1 5 - 6 1266 60
XM_005615775.2 XP_005615832.1 6 - 7 1446 60
XM_005615776.3 XP_005615833.1 5 - 6 1345 60
GenBank mRNA DQ975208.1 - 5 - 6 1266 60
DQ975209.1 - 5 - 6 1266 60
DQ975210.1 - 3 - 4 732 60
DQ975211.1 - 3 - 4 732 60

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Canis familiaris Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline