This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf03054212_m1
See other CEACAM30 GE Assays ›
Gene Symbol:
CEACAM30 CEACAM28
Gene Name:
carcinoembryonic antigen-related cell adhesion molecule 30
carcinoembryonic antigen-related cell adhesion molecule 28
Gene Aliases:
-
CEACAM21, CEACAM30
Chromosome Location:
-
Chromosome Location:
Chr.1: 111812571 - 111834985 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
-
Assay ID Cf03054212_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CEACAM30
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001097553.1 NP_001091022.1 5 - 6 916 113
Gene Symbol: CEACAM28
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA EF137912.1 - 5 - 6 916 113

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Canis familiaris Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0