This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf04647891_s1
See other MIR132 GE Assays ›
Gene Symbol:
MIR132
Gene Name:
microRNA mir-132
Gene Aliases:
cfa-mir-132
Chromosome Location:
Chr.9: 46153531 - 46153590 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf04647891_s1
Size
Availability Made To Order
Catalog # 4426961
Price 4,565.00
Your Price
Online offer:
4565.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR132
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_049382.1 1 30 59

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream carbonic anhydrase 10 CA10 -100.8kb Sense Chr. 9 - 27382508 - 27863806 on NCBI Build CanFam3.1
Downstream kinesin family member 2B KIF2B 1.3mb Anti-sense Chr. 9 - 29220252 - 29226481 on NCBI Build CanFam3.1

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline