This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cf04947064_g1
See other RPL32 GE Assays ›
Gene Symbol:
RPL32
Gene Name:
ribosomal protein L32
Gene Aliases:
-
Chromosome Location:
Chr.20: 5727201 - 5756190 on Build CanFam3.1
Species:
Dog
Species Specific ID (Flybase ID):
-
Assay ID Cf04947064_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RPL32
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001252169.1 NP_001239098.1 1 42 95
XM_022406256.1 XP_022261964.1 1691 95
XM_022406257.1 XP_022261965.1 1616 95
GenBank mRNA BQ234354.1 - 1 51 95

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001252169.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

calmodulin-related ribosomal protein translational protein

Gene Ontology Categories:

Function(s) Process(es)

translation
structural constituent of ribosome

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline