This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cg01218202_g1
See other CHTOP GE Assays ›
Gene Symbol:
Chtop
Gene Name:
chromatin target of PRMT1
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_003613854: 1689061 - 1697859 on Build CriGri-PICRH-1.0
Species:
Cricetulus griseus
Species Specific ID (Flybase ID):
-
Assay ID Cg01218202_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 417.00
Your Price
Online offer: 387.65
387.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Chtop
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003501636.2 XP_003501684.2 1 - 2 104 67
XM_007642298.1 XP_007640488.1 1 - 2 104 67
XM_007642299.1 XP_007640489.1 1 - 2 104 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

RNA metabolism protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-774fdbf798-5bs74:80/100.66.73.235:80.
git-commit: 7dd85a2aaed5e3122cf027567276b2918d4b9fce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.0-Offline