Assay Details
| Gene Symbol: Ppp2r5e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499304.2 | XP_003499352.1 | 10 - 11 | 1112 | 116 | |
| GenBank mRNA | JI875905.1 | - | 10 - 11 | 1112 | 116 | |
| JP053836.1 | - | 10 - 11 | 1112 | 116 | ||
More Information
Set Membership: |
|

Ppp2r5e
Genomic Map