This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cg04423246_m1
See other GMNN GE Assays ›
Gene Symbol:
Gmnn
Gene Name:
geminin, DNA replication inhibitor
Gene Aliases:
I79_008728
Chromosome Location:
Chr.Un|NW_003613881: 1404207 - 1414643 on Build CriGri-PICRH-1.0
Species:
Cricetulus griseus
Species Specific ID (Flybase ID):
-
Assay ID Cg04423246_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gmnn
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001246817.1 NP_001233746.1 5 - 6 460 72
XM_007642790.1 XP_007640980.1 6 - 7 610 72
XM_007642791.1 XP_007640981.1 6 - 7 612 72
XM_007642792.1 XP_007640982.1 6 - 7 757 72
GenBank mRNA AM503076.1 - 5 - 6 460 72
JI866002.1 - 5 - 6 469 72
JP053833.1 - 5 - 6 467 72

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0