This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cg04428497_m1
See other MMP28 GE Assays ›
Gene Symbol:
Mmp28
Gene Name:
matrix metallopeptidase 28
Gene Aliases:
I79_019777
Chromosome Location:
Chr.Un|NW_003615071: 104626 - 127480 on Build CriGri-PICRH-1.0
Species:
Cricetulus griseus
Species Specific ID (Flybase ID):
-
Assay ID Cg04428497_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mmp28
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_007651829.1 XP_007650019.1 5 - 6 1032 64
XM_007651830.1 XP_007650020.1 4 - 5 807 64

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Protease Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

metalloprotease

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-gqkn5:80/100.66.76.113:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0