Assay Details
| Gene Symbol: LOC100756144 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007643158.1 | XP_007641348.1 | 28 - 29 | 3488 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100756144| Gene Symbol: LOC100756144 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007643158.1 | XP_007641348.1 | 28 - 29 | 3488 | 74 | |
Set Membership: |
|