This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cg04466071_m1
See other DCTPP1 GE Assays ›
Gene Symbol:
Dctpp1
Gene Name:
dCTP pyrophosphatase 1
Gene Aliases:
I79_024463
Chromosome Location:
Chr.Un|NW_003617301: 6714 - 10149 on Build CriGri-PICRH-1.0
Species:
Cricetulus griseus
Species Specific ID (Flybase ID):
-
Assay ID Cg04466071_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dctpp1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003515071.2 XP_003515119.1 2 - 3 283 76
GenBank mRNA JI886586.1 - 2 - 3 274 76

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

hydrolase phosphatase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0