This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cg04490033_m1
See other ELAVL2 GE Assays ›
Gene Symbol:
Elavl2
Gene Name:
ELAV like neuron-specific RNA binding protein 2
Gene Aliases:
I79_015657
Chromosome Location:
Chr.Un|NW_003614445: 323301 - 454438 on Build CriGri-PICRH-1.0
Species:
Cricetulus griseus
Species Specific ID (Flybase ID):
-
Assay ID Cg04490033_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Elavl2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003507901.2 XP_003507949.2 2 - 3 316 81
XM_007648472.1 XP_007646662.1 2 - 3 316 81
XM_007648473.1 XP_007646663.1 2 - 3 316 81
XM_007648474.1 XP_007646664.1 2 - 3 315 81
XM_007648476.1 XP_007646666.1 2 - 3 781 81
XM_007648477.1 XP_007646667.1 2 - 3 273 81

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

translation initiation factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vh84l:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0